Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000098.2 | NP_000089.1 | 4 | 1421 | ||
XM_005270484.1 | XP_005270541.1 | 4 | 1089 | |||
GenBank mRNA | AK223052.1 | - | 2 | 664 | ||
AK312687.1 | - | 4 | 1025 | |||
BC002445.2 | - | 4 | 1030 | 3346025 | ||
BC005172.2 | - | 4 | 1030 | 3605012 | ||
M58581.1 | - | 4 | 1007 | |||
U09648.1 | - | 4 | 1422 |