Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000573.3 | NP_000564.2 | 10 | 1742 | ||
NM_000651.4 | NP_000642.3 | 10 | 1742 | |||
XM_006711166.2 | XP_006711229.1 | 10 | 1718 | |||
XM_011509205.1 | XP_011507507.1 | 10 | 1718 | |||
GenBank mRNA | AK309342.1 | - | 10 | 1723 | ||
BC032550.1 | - | 10 | 1631 | |||
BX643705.1 | - | 5 | 574 | |||
X05309.1 | - | 1 | 98 | |||
X14362.1 | - | 10 | 1529 | |||
Y00816.1 | - | 1629 |