Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000573.3 | NP_000564.2 | 27 | 4538 | ||
NM_000651.4 | NP_000642.3 | 35 | 5888 | |||
XM_006711166.2 | XP_006711229.1 | 35 | 5864 | |||
XM_011509205.1 | XP_011507507.1 | 35 | 5864 | |||
GenBank mRNA | BC032550.1 | - | 19 | 3077 | ||
X05309.1 | - | 18 | 2894 | |||
Y00816.1 | - | 4425 |