Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRABP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004378.2 | NP_004369.1 | 1 | 153 | ||
GenBank mRNA | AL110113.1 | - | 1 | 37 | ||
BC022069.1 | - | 1 | 129 | 4797853 |