Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005206.4 | NP_005197.3 | 2 | 490 | ||
NM_016823.3 | NP_058431.2 | 2 | 490 | |||
GenBank mRNA | AK291060.1 | - | 2 | 455 | ||
AU125411.1 | - | 2 | 490 | |||
BC001718.1 | - | 2 | 442 | 3353237 | ||
BC008506.1 | - | 2 | 473 | 3452137 | ||
BC009837.1 | - | 2 | 455 | 4040536 | ||
BT007277.1 | - | 2 | 340 | |||
D10656.1 | - | 2 | 445 | |||
KU177996.1 | - | 2 | 340 | |||
KU177997.1 | - | 2 | 340 |