Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASB11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012428.2 | NP_001012428.1 | 3 | 348 | ||
NM_001201583.1 | NP_001188512.1 | 3 | 306 | |||
NM_080873.2 | NP_543149.1 | 3 | 357 | |||
GenBank mRNA | AF425642.1 | - | 3 | 338 | ||
BC069340.1 | - | 3 | 338 | 7262279 | ||
BC103873.1 | - | 3 | 338 | |||
BC103874.1 | - | 3 | 338 | 40004166 | ||
BC103875.1 | - | 3 | 338 | 40004167 | ||
BX537853.1 | - | 3 | 348 | |||
BX537857.1 | - | 3 | 306 |