Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASB8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001319296.1 | NP_001306225.1 | 3 | 416 | ||
NM_001319297.1 | NP_001306226.1 | 2 | 276 | |||
NM_001319298.1 | NP_001306227.1 | 2 | 280 | |||
NM_001319299.1 | NP_001306228.1 | 2 | 276 | |||
NM_001319300.1 | NP_001306229.1 | 2 | 280 | |||
NM_001319301.1 | NP_001306230.1 | 2 | 276 | |||
NM_024095.4 | NP_077000.1 | 2 | 280 | |||
GenBank mRNA | AF464877.1 | - | 2 | 172 | ||
AK024908.1 | - | 2 | 181 | |||
AK289957.1 | - | 2 | 247 | |||
AK297013.1 | - | 2 | 185 | |||
BC001321.1 | - | 2 | 187 | 3456049 | ||
BM837218.1 | - | 2 | 280 | |||
BP195070.1 | - | 2 | 276 | |||
BP210910.1 | - | 2 | 245 | |||
BP372866.1 | - | 2 | 276 | |||
CR457336.1 | - | 1 | 110 | |||
DA071750.1 | - | 3 | 338 | |||
DA979010.1 | - | 2 | 197 |