Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHODL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204174.1 | NP_001191103.1 | 2 | 200 | ||
NM_001204175.1 | NP_001191104.1 | 2 | 166 | |||
NM_001204176.1 | NP_001191105.1 | 3 | 266 | |||
NM_001204177.1 | NP_001191106.1 | 2 | 166 | |||
NM_001204178.1 | NP_001191107.1 | 3 | 266 | |||
NM_024944.2 | NP_079220.2 | 2 | 575 | |||
XM_011529453.2 | XP_011527755.1 | 2 | 809 | |||
XM_011529457.2 | XP_011527759.1 | 3 | 572 | |||
XM_017028273.1 | XP_016883762.1 | 2 | 195 | |||
GenBank mRNA | AF257472.1 | - | 2 | 575 | ||
AF523313.1 | - | 3 | 265 | |||
AF523314.1 | - | 3 | 266 | |||
AF523315.1 | - | 2 | 167 | |||
AK022689.1 | - | 3 | 448 | |||
AK296009.1 | - | 2 | 200 | |||
AK313725.1 | - | 2 | 223 | |||
AY358608.1 | - | 2 | 561 | |||
BC009418.1 | - | 2 | 545 | 3509743 |