Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WFDC6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080827.1 | NP_543017.1 | 2, 3 | 309 | ||
GenBank mRNA | AF411861.1 | - | 2, 3 | 309 | ||
BC101600.1 | - | 2, 3 | 304 | 8069106 | ||
BC101604.1 | - | 2, 3 | 304 | 8069110 | ||
GQ472217.1 | - | 2, 3 | 304 |