Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SIRPA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040022.1 | NP_001035111.1 | 6, 7 | 1564 | ||
NM_001040023.1 | NP_001035112.1 | 5, 6 | 1472 | |||
NM_080792.2 | NP_542970.1 | 6, 7 | 1231 | |||
XM_005260669.3 | XP_005260726.1 | 6, 7 | 1614 | |||
XM_005260670.3 | XP_005260727.1 | 6, 7 | 1230 | |||
XM_006723545.3 | XP_006723608.1 | 3, 4 | 801 | |||
XM_011529173.2 | XP_011527475.1 | 5, 6 | 1219 | |||
GenBank mRNA | AB023430.1 | - | 6, 7 | 1231 | ||
AK290776.1 | - | 6, 7 | 1337 | |||
AK298243.1 | - | 6, 7 | 1182 | |||
AK312521.1 | - | 5, 6 | 1285 | |||
BC026692.1 | - | 6, 7 | 1236 | 3916836 | ||
BC033092.1 | - | 5, 6 | 1224 | 4589298 | ||
BC038510.2 | - | 5, 6 | 1469 | 5261789 | ||
BC075849.1 | - | 6, 7 | 1410 | 4589732 | ||
D86043.1 | - | 5, 6 | 1203 | |||
U06701.1 | - | 4, 5 | 1131 | |||
Y10375.1 | - | 5, 6 | 1213 | |||
Y11047.1 | - | 4, 5 | 979 |