Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASB12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130388.3 | NP_569059.3 | 2 | 909 | ||
GenBank mRNA | AF403030.1 | - | 1 | 689 | ||
AK096896.1 | - | 2 | 889 | |||
AK300684.1 | - | 3 | 789 | |||
BC069436.1 | - | 1 | 689 | 7262244 | ||
BC069555.1 | - | 1 | 689 | 7262256 | ||
BC093883.1 | - | 1 | 750 | 7939728 | ||
BC104946.1 | - | 1 | 750 | 8143949 |