Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C10orf82 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144661.2 | NP_653262.1 | 4 | 427 | ||
XM_011539340.2 | XP_011537642.1 | 4 | 882 | |||
XM_011539341.1 | XP_011537643.1 | 4 | 466 | |||
XM_011539342.2 | XP_011537644.1 | 4 | 593 | |||
XM_011539343.2 | XP_011537645.1 | 4 | 878 | |||
XM_011539344.2 | XP_011537646.1 | 4 | 845 | |||
XM_011539345.2 | XP_011537647.1 | 4 | 849 | |||
XM_011539346.2 | XP_011537648.1 | 4 | 849 | |||
XM_011539347.2 | XP_011537649.1 | 4 | 864 | |||
XM_017015753.1 | XP_016871242.1 | 4 | 466 | |||
XM_017015754.1 | XP_016871243.1 | 4 | 592 | |||
XM_017015755.1 | XP_016871244.1 | 4 | 608 | |||
GenBank mRNA | AK097587.1 | - | 3 | 1078 | ||
BC021737.2 | - | 4 | 427 | 4821913 | ||
BX537582.1 | - | 3 | 1228 |