Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CSF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000757.5 | NP_000748.3 | 8 | 2063 | ||
NM_172210.2 | NP_757349.1 | 9 | 1715 | |||
NM_172211.3 | NP_757350.1 | 8 | 1169 | |||
NM_172212.2 | NP_757351.1 | 8 | 2063 | |||
XM_017000369.1 | XP_016855858.1 | 8 | 1738 | |||
GenBank mRNA | AB208952.1 | - | 8 | 1813 | ||
AK291660.1 | - | 8 | 1825 | |||
AK300401.1 | - | 6 | 1445 | |||
AK311440.1 | - | 5 | 461 | |||
BC021117.1 | - | 8 | 1839 | 4586312 | ||
M27087.1 | - | 8 | 1771 | |||
M37435.1 | - | 7 | 1755 | |||
M64592.1 | - | 8 | 1650 | |||
M76453.1 | - | 7 | 720 | |||
U22386.1 | - | 9 | 1346 | |||
X05825.1 | - | 7 | 1598 |