Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARHGAP42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152432.2 | NP_689645.2 | 6 | 529 | ||
XM_011542615.2 | XP_011540917.1 | 6 | 3017 | |||
XM_011542616.2 | XP_011540918.1 | 6 | 786 | |||
XM_011542617.2 | XP_011540919.1 | 7 | 672 | |||
XM_017017237.1 | XP_016872726.1 | 6 | 975 | |||
XM_017017238.1 | XP_016872727.1 | 7 | 3158 | |||
GenBank mRNA | DA800121.1 | - | 2 | 94 |