Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPNE8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153634.2 | NP_705898.1 | 19, 20 | 1598 | ||
XM_017018851.1 | XP_016874340.1 | 20, 21 | 2571 | |||
XM_017018852.1 | XP_016874341.1 | 19, 20 | 1544 | |||
GenBank mRNA | AK074223.1 | - | 5, 6 | 460 | ||
AK094998.1 | - | 7, 8 | 1172 | |||
AK098593.1 | - | 19, 20 | 1529 | |||
AK307908.1 | - | 19, 20 | 1569 | |||
AY177785.1 | - | 19, 20 | 1514 | |||
BC046366.1 | - | 7, 8 | 915 | |||
BC110577.1 | - | 19, 20 | 1589 | 40007743 | ||
BC110578.1 | - | 19, 20 | 1589 | 40007744 | ||
BX538055.1 | - | 19, 20 | 1612 |