Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KRT80 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081492.1 | NP_001074961.1 | 6 | 993 | ||
NM_182507.2 | NP_872313.2 | 6 | 993 | |||
XM_005268676.3 | XP_005268733.1 | 4 | 1088 | |||
GenBank mRNA | AJ717743.1 | - | 6 | 947 | ||
BC065180.1 | - | 6 | 993 | 6573884 | ||
BU151185.1 | - | 3 | 295 |