Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SERPINA12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304461.1 | NP_001291390.1 | 5 | 1450 | ||
NM_173850.3 | NP_776249.1 | 6 | 2023 | |||
XM_011536451.2 | XP_011534753.1 | 5 | 1426 | |||
XM_011536452.2 | XP_011534754.1 | 5 | 1420 | |||
XM_011536453.2 | XP_011534755.1 | 5 | 1332 | |||
XM_011536454.2 | XP_011534756.1 | 5 | 1411 | |||
XM_017020989.1 | XP_016876478.1 | 5 | 1421 | |||
XM_017020990.1 | XP_016876479.1 | 5 | 1406 | |||
GenBank mRNA | AY177692.1 | - | 4 | 1245 | ||
AY326420.1 | - | 5 | 1450 | |||
BC040857.1 | - | 6 | 2023 | 5527002 |