Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CSNK2A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001895.3 | NP_001886.1 | 12 | 1317 | ||
NM_177559.2 | NP_808227.1 | 13 | 1434 | |||
NM_177560.2 | NP_808228.1 | 11 | 1107 | |||
GenBank mRNA | AB451279.1 | - | 11 | 1041 | ||
AB451411.1 | - | 11 | 1041 | |||
AK302583.1 | - | 11 | 996 | |||
AK312535.1 | - | 13 | 1325 | |||
BC011668.2 | - | 12 | 1189 | 4121882 | ||
BC050036.1 | - | 12 | 1208 | 5298655 | ||
BC053532.1 | - | 11 | 1136 | 3908058 | ||
BC071167.1 | - | 11 | 1048 | 5772615 | ||
BT019792.1 | - | 11 | 1041 | |||
DQ354583.1 | - | 12 | 1183 | |||
J02853.1 | - | 12 | 1194 | |||
M55265.1 | - | 12 | 1189 | |||
S53149.1 | - | 12 | 1194 |