Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MESP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039958.1 | NP_001035047.1 | 2 | 1180 | ||
GenBank mRNA | AL360139.1 | - | 3 | 425 | ||
BC111413.1 | - | 2 | 1168 | 40004357 |