Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZSCAN29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152455.3 | NP_689668.3 | 3 | 796 | ||
XM_006720401.3 | XP_006720464.1 | 4 | 1556 | |||
XM_011521266.2 | XP_011519568.1 | 4 | 1236 | |||
XM_017021949.1 | XP_016877438.1 | 3 | 1768 | |||
GenBank mRNA | AF525399.1 | - | 3 | 646 | ||
AK093186.1 | - | 2 | 342 | |||
AK122788.1 | - | 3 | 784 | |||
BC043612.1 | - | 3 | 764 | |||
BC062551.1 | - | 3 | 412 | |||
BC109270.1 | - | 3 | 646 | 40026738 |