Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NCAN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004386.2 | NP_004377.2 | 11 | 3494 | ||
GenBank mRNA | AB210004.1 | - | 11 | 3438 | ||
AF026547.1 | - | 10 | 3396 | |||
AK297395.1 | - | 5 | 1800 | |||
AK309138.1 | - | 5 | 1966 |