Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172771.1 | NP_001166242.1 | 6, 7 | 1097 | ||
NM_001172772.1 | NP_001166243.1 | 6, 7 | 1083 | |||
NM_152456.2 | NP_689669.2 | 6, 7 | 1100 | |||
XM_011522902.2 | XP_011521204.1 | 6, 7 | 1184 | |||
XM_011522903.2 | XP_011521205.1 | 6, 7 | 966 | |||
GenBank mRNA | AK172744.1 | - | 5, 6 | 615 | ||
AK307727.1 | - | 5, 6 | 735 | |||
AK314913.1 | - | 5, 6 | 620 | |||
BC029804.1 | - | 5, 6 | 928 | 5192904 | ||
EU599219.1 | - | 5, 6 | 542 | |||
EU636994.1 | - | 5, 6 | 545 |