Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRSS36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001258290.1 | NP_001245219.1 | 6 | 713 | ||
NM_001258291.1 | NP_001245220.1 | 6 | 713 | |||
NM_173502.4 | NP_775773.2 | 6 | 713 | |||
XM_011545758.2 | XP_011544060.1 | 5 | 931 | |||
XM_017022968.1 | XP_016878457.1 | 6 | 702 | |||
XM_017022969.1 | XP_016878458.1 | 6 | 702 | |||
XM_017022970.1 | XP_016878459.1 | 6 | 702 | |||
XM_017022971.1 | XP_016878460.1 | 6 | 702 | |||
GenBank mRNA | AJ627034.1 | - | 6 | 685 | ||
AK075142.1 | - | 6 | 686 | |||
AK290310.1 | - | 6 | 713 | |||
AK297997.1 | - | 4 | 654 | |||
AK301409.1 | - | 6 | 685 | |||
AW450407.1 | - | 2 | 104 | |||
BC137396.1 | - | 6 | 680 | 9021394 | ||
BC144615.1 | - | 6 | 680 | 9053146 |