Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CSRP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300965.1 | NP_001287894.1 | 2, 3 | 183 | ||
NM_001321.2 | NP_001312.1 | 2, 3 | 208 | |||
GenBank mRNA | BC000992.2 | - | 2, 3 | 156 | 3448693 | |
BT019913.1 | - | 1, 2 | 104 | |||
BX461295.2 | - | 2, 3 | 230 | |||
CN408330.1 | - | 2, 3 | 154 | |||
U46006.1 | - | 1, 2 | 104 | |||
U57646.1 | - | 2, 3 | 160 |