Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC47A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099646.1 | NP_001093116.1 | 13 | 1312 | ||
NM_001256663.1 | NP_001243592.1 | 14 | 1354 | |||
NM_152908.3 | NP_690872.2 | 13 | 1420 | |||
NR_135624.1 | 12 | 1807 | ||||
NR_135625.1 | 12 | 1390 | ||||
XM_017024221.1 | XP_016879710.1 | 14 | 1396 | |||
XM_017024222.1 | XP_016879711.1 | 14 | 1896 | |||
XM_017024223.1 | XP_016879712.1 | 14 | 1396 | |||
XM_017024224.1 | XP_016879713.1 | 14 | 1400 | |||
GenBank mRNA | AB250364.1 | - | 13 | 1155 | ||
AB250701.1 | - | 12 | 1233 | |||
AK055758.1 | - | 13 | 1198 | |||
BC035288.1 | - | 14 | 1222 | 5189385 | ||
BC050578.1 | - | 13 | 1296 | 5759988 | ||
BX648861.1 | - | 12 | 1807 |