Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EME1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166131.1 | NP_001159603.1 | 8 | 1593 | ||
NM_152463.2 | NP_689676.2 | 8 | 1554 | |||
XM_005257081.3 | XP_005257138.1 | 8 | 1079 | |||
XM_011524395.2 | XP_011522697.1 | 8 | 1044 | |||
XM_017024236.1 | XP_016879725.1 | 7 | 1467 | |||
GenBank mRNA | AK021607.1 | - | 1 | 679 | ||
AK055926.1 | - | 8 | 1545 | |||
BC016470.2 | - | 7 | 1536 | 2899969 |