Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF418 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317027.1 | NP_001303956.1 | 5 | 1003 | ||
NM_001317028.1 | NP_001303957.1 | 5 | 717 | |||
NM_001317029.1 | NP_001303958.1 | 5 | 665 | |||
NM_001317030.1 | NP_001303959.1 | 2 | 570 | |||
NM_133460.2 | NP_597717.1 | 4 | 783 | |||
XM_017026304.1 | XP_016881793.1 | 5 | 1020 | |||
XM_017026305.1 | XP_016881794.1 | 5 | 1020 | |||
XM_017026306.1 | XP_016881795.1 | 5 | 1017 | |||
XM_017026307.1 | XP_016881796.1 | 3 | 715 | |||
XM_017026308.1 | XP_016881797.1 | 3 | 715 | |||
XM_017026309.1 | XP_016881798.1 | 4 | 782 | |||
XM_017026310.1 | XP_016881799.1 | 5 | 735 | |||
XM_017026311.1 | XP_016881800.1 | 5 | 735 | |||
XM_017026312.1 | XP_016881801.1 | 4 | 783 | |||
XM_017026313.1 | XP_016881802.1 | 5 | 1758 | |||
XM_017026314.1 | XP_016881803.1 | 4 | 645 | |||
GenBank mRNA | AB075836.1 | - | 3 | 617 | ||
AK056113.1 | - | 4 | 770 | |||
AK293723.1 | - | 5 | 1003 | |||
AK304632.1 | - | 2 | 564 | |||
AY695825.1 | - | 4 | 768 | |||
BC112244.1 | - | 3 | 567 | 8327712 | ||
BC113707.1 | - | 3 | 567 | 8322759 |