Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PROSER3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039887.2 | NP_001034976.2 | 3 | 278 | ||
XM_011526528.1 | XP_011524830.1 | 3 | 268 | |||
XM_011526529.2 | XP_011524831.1 | 3 | 282 | |||
XM_011526530.1 | XP_011524832.1 | 3 | 268 | |||
XM_011526531.1 | XP_011524833.1 | 3 | 268 | |||
XM_011526532.1 | XP_011524834.1 | 3 | 268 | |||
XM_011526533.2 | XP_011524835.1 | 3 | 268 | |||
XM_011526534.1 | XP_011524836.1 | 3 | 268 | |||
XM_011526536.2 | XP_011524838.1 | 3 | 282 | |||
GenBank mRNA | AK055219.1 | - | 3 | 237 | ||
AK124144.1 | - | 1 | 1687 | |||
AL833894.1 | - | 3 | 242 | |||
BC017947.1 | - | 3 | 227 | |||
BC110893.1 | - | 2 | 209 | 3914279 | ||
DB086668.1 | - | 3 | 270 |