Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM56 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199679.1 | NP_001186608.1 | 7 | 949 | ||
NM_152487.2 | NP_689700.1 | 7 | 864 | |||
GenBank mRNA | AK056404.1 | - | 7 | 774 | ||
BC137457.1 | - | 7 | 664 | 9021457 | ||
BC137458.1 | - | 7 | 664 | 9021458 | ||
BC139737.1 | - | 7 | 664 | 40053604 |