Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CTBS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004388.2 | NP_004379.1 | 6 | 996 | ||
GenBank mRNA | AK075423.1 | - | 6 | 1260 | ||
AK298703.1 | - | 6 | 947 | |||
AK314640.1 | - | 6 | 962 | |||
BC024007.2 | - | 5 | 778 | |||
BC096752.1 | - | 5 | 778 | |||
BC126333.1 | - | 6 | 972 | 8992049 | ||
BC126335.1 | - | 6 | 972 | 8992051 | ||
M95767.1 | - | 6 | 931 |