Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CTBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012614.1 | NP_001012632.1 | 6 | 948 | ||
NM_001328.2 | NP_001319.1 | 5 | 753 | |||
XM_005272261.3 | XP_005272318.1 | 5 | 764 | |||
XM_005272263.4 | XP_005272320.1 | 6 | 6979 | |||
XM_011513395.2 | XP_011511697.1 | 6 | 808 | |||
XM_011513396.2 | XP_011511698.1 | 6 | 913 | |||
XM_011513397.2 | XP_011511699.1 | 5 | 6696 | |||
XM_017007762.1 | XP_016863251.1 | 6 | 962 | |||
XM_017007763.1 | XP_016863252.1 | 6 | 1085 | |||
XM_017007764.1 | XP_016863253.1 | 6 | 6979 | |||
XM_017007765.1 | XP_016863254.1 | 6 | 808 | |||
XM_017007766.1 | XP_016863255.1 | 6 | 913 | |||
XM_017007767.1 | XP_016863256.1 | 6 | 827 | |||
GenBank mRNA | AF091555.1 | - | 5 | 634 | ||
BC011655.2 | - | 5 | 626 | 4128336 | ||
BC053320.1 | - | 6 | 940 | 6461670 | ||
BC064333.1 | - | 6 | 2346 | |||
BC098400.1 | - | 5 | 763 | 30344174 | ||
KJ534818.1 | - | 5 | 588 | |||
KJ535006.1 | - | 5 | 588 | |||
KJ535043.1 | - | 4 | 237 | |||
KJ535050.1 | - | 4 | 433 | |||
KJ535065.1 | - | 6 | 783 | |||
KJ535066.1 | - | 6 | 783 | |||
KJ535067.1 | - | 6 | 783 | |||
U37408.1 | - | 5 | 637 |
Gene Symbol: CTBP1-AS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_104331.1 | 6 | 1456 | |||
GenBank mRNA | AK092548.1 | - | 6 | 1456 | ||
AX747592.1 | - | 6 | 1456 |