Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PM20D1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152491.4 | NP_689704.4 | 1 | 115 | ||
NR_135186.1 | 1 | 115 | ||||
XM_017000396.1 | XP_016855885.1 | 1 | 109 | |||
XM_017000397.1 | XP_016855886.1 | 1 | 109 | |||
XM_017000398.1 | XP_016855887.1 | 1 | 109 | |||
GenBank mRNA | AK057131.1 | - | 1 | 99 | ||
AK290786.1 | - | 1 | 99 | |||
BC039170.1 | - | 1 | 84 | 4746689 | ||
BC063477.1 | - | 1 | 84 | 5221187 | ||
BI832659.1 | - | 1 | 84 | |||
DA387977.1 | - | 1 | 115 |