Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLIS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_147193.2 | NP_671726.2 | 9 | 2161 | ||
XM_017000408.1 | XP_016855897.1 | 10 | 2228 | |||
XM_017000409.1 | XP_016855898.1 | 10 | 2216 | |||
XM_017000410.1 | XP_016855899.1 | 10 | 2342 | |||
GenBank mRNA | AK090634.1 | - | 9 | 2160 | ||
AK093474.1 | - | 7 | 1530 | |||
BC101799.1 | - | 7 | 1667 | 8069305 | ||
BC104911.1 | - | 7 | 1667 | 8143914 |