Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF362 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152493.2 | NP_689706.2 | 3, 4 | 278 | ||
XM_005270507.2 | XP_005270564.1 | 2, 3 | 418 | |||
XM_005270508.4 | XP_005270565.1 | 3, 4 | 1075 | |||
XM_017000415.1 | XP_016855904.1 | 2, 3 | 990 | |||
GenBank mRNA | AF318356.1 | - | 2, 3 | 128 | ||
BC104779.1 | - | 2, 3 | 200 | 8143782 | ||
BC112113.1 | - | 2, 3 | 200 | 8327581 | ||
BX283663.1 | - | 3, 4 | 279 |