Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CTLA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037631.2 | NP_001032720.1 | 2 | 479 | ||
NM_005214.4 | NP_005205.2 | 2 | 479 | |||
GenBank mRNA | AF414120.1 | - | 2 | 479 | ||
AF486806.1 | - | 2 | 324 | |||
AK313732.1 | - | 2 | 464 | |||
AY209009.1 | - | 2 | 324 | |||
BC069566.1 | - | 2 | 419 | 7262243 | ||
BC070162.1 | - | 2 | 480 | 30417685 | ||
BC074842.2 | - | 2 | 350 | 30915552 | ||
BC074893.2 | - | 2 | 355 | 30915247 | ||
CD639535.1 | - | 2 | 483 | |||
L15006.1 | - | 2 | 324 | |||
U90273.1 | - | 1 | 213 |