Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C22orf15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182520.2 | NP_872326.2 | 4 | 562 | ||
XM_011529907.2 | XP_011528209.1 | 3 | 1353 | |||
XM_011529908.2 | XP_011528210.2 | 4 | 1074 | |||
XM_011529912.2 | XP_011528214.1 | 2 | 1194 | |||
XM_017028602.1 | XP_016884091.1 | 3 | 1358 | |||
XM_017028603.1 | XP_016884092.1 | 3 | 1337 | |||
XM_017028604.1 | XP_016884093.1 | 3 | 1358 | |||
XM_017028605.1 | XP_016884094.1 | 3 | 1368 | |||
XM_017028606.1 | XP_016884095.1 | 3 | 1358 | |||
XM_017028607.1 | XP_016884096.1 | 3 | 1358 | |||
XM_017028608.1 | XP_016884097.1 | 3 | 1358 | |||
XM_017028609.1 | XP_016884098.1 | 4 | 1087 | |||
XM_017028610.1 | XP_016884099.1 | 4 | 1065 | |||
XM_017028611.1 | XP_016884100.1 | 3 | 1358 | |||
XM_017028612.1 | XP_016884101.1 | 4 | 1058 | |||
XM_017028613.1 | XP_016884102.1 | 3 | 1368 | |||
XM_017028614.1 | XP_016884103.1 | 3 | 1353 | |||
XM_017028615.1 | XP_016884104.1 | 4 | 1064 | |||
XM_017028616.1 | XP_016884105.1 | 4 | 1075 | |||
XM_017028617.1 | XP_016884106.1 | 4 | 1066 | |||
GenBank mRNA | AK093880.1 | - | 3 | 824 | ||
BC113450.1 | - | 1 | 186 | 8322502 | ||
BC113452.1 | - | 1 | 186 | 8322504 | ||
BU739625.1 | - | 3 | 314 | |||
CR456371.1 | - | 4 | 375 |