Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HORMAD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011529914.2 | XP_011528216.1 | 2 | 480 | ||
XM_011529917.2 | XP_011528219.1 | 2 | 339 | |||
XM_011529918.1 | XP_011528220.1 | 2 | 480 | |||
XM_017028621.1 | XP_016884110.1 | 2 | 361 | |||
XM_017028622.1 | XP_016884111.1 | 2 | 280 | |||
XM_017028624.1 | XP_016884113.1 | 2 | 480 | |||
GenBank mRNA | AK098703.1 | - | 2 | 305 | ||
BC030013.1 | - | 2 | 90 | 4823215 | ||
BI459310.1 | - | 2 | 85 | |||
CR456341.1 | - | 1 | 67 | |||
HY002382.1 | - | 3 | 130 |