Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C22orf42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010859.1 | NP_001010859.1 | 7 | 561 | ||
XM_011529922.2 | XP_011528224.1 | 8 | 1060 | |||
XM_017028629.1 | XP_016884118.1 | 8 | 594 | |||
XM_017028630.1 | XP_016884119.1 | 6 | 970 | |||
XM_017028631.1 | XP_016884120.1 | 8 | 1140 | |||
GenBank mRNA | CR456485.1 | - | 7 | 561 |