Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAJB7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145174.1 | NP_660157.1 | 1 | 204 | ||
GenBank mRNA | AF085232.1 | - | 1 | 204 | ||
BC112135.1 | - | 1 | 163 | 8327603 | ||
CR456468.1 | - | 1 | 109 |
Gene Symbol: XPNPEP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204827.1 | NP_001191756.1 | 3 | 583 | ||
GenBank mRNA | BC005331.1 | - | 3 | 567 |