Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CTPS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301237.1 | NP_001288166.1 | 5 | 597 | ||
NM_001905.3 | NP_001896.2 | 9 | 1433 | |||
NR_125440.1 | 9 | 1433 | ||||
GenBank mRNA | AK130549.1 | - | 8 | 1019 | ||
AK225899.1 | - | 9 | 937 | |||
AK297386.1 | - | 9 | 1029 | |||
AK299122.1 | - | 5 | 409 | |||
AK303797.1 | - | 5 | 526 | |||
BC009408.1 | - | 9 | 1040 | 3355881 | ||
X52142.1 | - | 9 | 964 |