Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CTSE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001910.3 | NP_001901.1 | 2, 3 | 335 | ||
NM_148964.2 | NP_683865.1 | 2, 3 | 335 | |||
XM_011509244.1 | XP_011507546.1 | 2, 3 | 325 | |||
XM_011509245.1 | XP_011507547.1 | 2, 3 | 325 | |||
GenBank mRNA | AJ250716.1 | - | 2, 3 | 321 | ||
AJ250717.1 | - | 2, 3 | 321 | |||
AK292057.1 | - | 2, 3 | 265 | |||
BC042537.1 | - | 2, 3 | 302 | 5174814 | ||
J05036.1 | - | 2, 3 | 266 |