Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDSUB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128212.2 | NP_001121684.1 | 3 | 789 | ||
NM_001128213.1 | NP_001121685.1 | 3 | 805 | |||
NM_001307994.1 | NP_001294923.1 | 3 | 789 | |||
NM_152528.2 | NP_689741.2 | 3 | 697 | |||
XM_005246355.2 | XP_005246412.2 | 3 | 783 | |||
XM_006712341.3 | XP_006712404.1 | 2 | 277 | |||
XM_011510753.2 | XP_011509055.1 | 3 | 749 | |||
XM_011510754.2 | XP_011509056.1 | 3 | 483 | |||
XM_017003481.1 | XP_016858970.1 | 3 | 783 | |||
XM_017003482.1 | XP_016858971.1 | 3 | 783 | |||
XM_017003483.1 | XP_016858972.1 | 3 | 698 | |||
GenBank mRNA | AK093494.1 | - | 3 | 697 | ||
AK129983.1 | - | 3 | 805 | |||
AL040518.1 | - | 2 | 173 | |||
BC029520.1 | - | 3 | 693 | 5301559 |