Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPR155 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033045.3 | NP_001028217.1 | 5 | 1321 | ||
NM_001267050.1 | NP_001253979.1 | 5 | 1218 | |||
NM_001267051.1 | NP_001253980.1 | 4 | 1172 | |||
NM_152529.6 | NP_689742.4 | 4 | 1172 | |||
XM_017003485.1 | XP_016858974.1 | 6 | 1363 | |||
XM_017003486.1 | XP_016858975.1 | 4 | 1224 | |||
XM_017003487.1 | XP_016858976.1 | 5 | 1225 | |||
GenBank mRNA | AK056381.1 | - | 2 | 420 | ||
AK091200.1 | - | 4 | 1113 | |||
AK307896.1 | - | 4 | 1164 | |||
AK315125.1 | - | 4 | 1113 | |||
AL834154.1 | - | 5 | 1270 | |||
BC028730.1 | - | 5 | 1267 | 4813561 | ||
BC035037.1 | - | 5 | 1229 | 5260361 | ||
BC036681.1 | - | 4 | 1118 | |||
BX537947.1 | - | 4 | 1165 |