Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EFHB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144715.3 | NP_653316.3 | 1 | 417 | ||
XM_005264889.1 | XP_005264946.1 | 1 | 417 | |||
XM_011533382.1 | XP_011531684.1 | 1 | 417 | |||
XM_017005742.1 | XP_016861231.1 | 1 | 417 | |||
GenBank mRNA | AK097644.1 | - | 1 | 419 | ||
BC028198.1 | - | 1 | 383 | 5209454 |