Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRC34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172779.1 | NP_001166250.1 | 10 | 1415 | ||
NM_153353.4 | NP_699184.2 | 9 | 1319 | |||
XM_006713508.3 | XP_006713571.1 | 10 | 1145 | |||
XM_011512442.2 | XP_011510744.1 | 10 | 1196 | |||
XM_017005746.1 | XP_016861235.1 | 10 | 1370 | |||
GenBank mRNA | AK095125.1 | - | 9 | 1762 | ||
AK130856.1 | - | 3 | 388 | |||
AK293661.1 | - | 10 | 1293 | |||
AK302466.1 | - | 10 | 1177 | |||
BC032841.2 | - | 9 | 1058 | 4826965 |