Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPNE9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308388.1 | NP_001295317.1 | 8 | 586 | ||
NM_153635.2 | NP_705899.2 | 8 | 567 | |||
XM_011533386.2 | XP_011531688.1 | 7 | 743 | |||
XM_011533388.2 | XP_011531690.1 | 7 | 743 | |||
XM_011533389.2 | XP_011531691.1 | 7 | 743 | |||
XM_017005747.1 | XP_016861236.1 | 8 | 389 | |||
GenBank mRNA | BC035735.1 | - | 8 | 567 | 5729888 | |
BC130375.1 | - | 8 | 438 | 40146406 | ||
BM545574.1 | - | 8 | 567 | |||
BX453546.2 | - | 9 | 635 |