Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CTSO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001334.2 | NP_001325.1 | 4 | 531 | ||
GenBank mRNA | AK300112.1 | - | 2 | 154 | ||
AK308323.1 | - | 4 | 537 | |||
AK312354.1 | - | 4 | 496 | |||
BC049206.1 | - | 4 | 484 | 5189657 | ||
BI834113.1 | - | 4 | 534 | |||
X77383.1 | - | 4 | 531 |