Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_174908.3 | NP_777568.1 | 5 | 1012 | ||
NM_178335.2 | NP_848018.1 | 5 | 1012 | |||
XM_011512460.1 | XP_011510762.1 | 5 | 1015 | |||
GenBank mRNA | AJ416916.2 | - | 5 | 1012 | ||
AJ557013.1 | - | 5 | 1012 | |||
BC065004.1 | - | 5 | 509 | 6049272 |