Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPICE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144718.3 | NP_653319.1 | 15 | 2555 | ||
XM_011512461.2 | XP_011510763.1 | 15 | 2312 | |||
XM_017005775.1 | XP_016861264.1 | 15 | 2312 | |||
XM_017005776.1 | XP_016861265.1 | 15 | 2449 | |||
XM_017005777.1 | XP_016861266.1 | 15 | 2372 | |||
XM_017005778.1 | XP_016861267.1 | 15 | 2556 | |||
GenBank mRNA | AK126897.1 | - | 11 | 1961 | ||
AK129575.1 | - | 5 | 1084 | |||
AL832148.1 | - | 15 | 2524 | |||
AL832327.2 | - | 15 | 4220 | |||
AY099107.1 | - | 15 | 2475 | |||
BC019232.1 | - | 1 | 51 | |||
BC036951.1 | - | 15 | 2496 | 5500345 | ||
BC038595.1 | - | 3 | 678 |