Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SH3D19 SIRNA Assays ›
Gene Symbol:
SH3D19
Gene Name:
SH3 domain containing 19
Gene Aliases:
EBP, EVE1, Kryn, SH3P19
Chromosome Location:
Chr.4: 151120281 - 151228030 on Build GRCh38
UniGene ID:
Hs.744983
Species:
Human
siRNA ID
HSS175727
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SH3D19
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001009555.3
NP_001009555.3
15, 16
2737
NM_001128923.1
NP_001122395.1
14, 15
2656
NM_001128924.1
NP_001122396.1
7, 8
1659
NM_001243349.1
NP_001230278.1
14, 15
2577
XM_005262767.2
XP_005262824.1
16, 17
2598
XM_011531646.1
XP_011529948.1
14, 15
2384
XM_011531647.1
XP_011529949.1
13, 14
2276
XM_011531648.1
XP_011529950.1
16, 17
2711
XM_011531649.1
XP_011529951.1
12, 13
2207
XM_017007778.1
XP_016863267.1
16, 17
2702
XM_017007779.1
XP_016863268.1
12, 13
2198
XM_017007780.1
XP_016863269.1
15, 16
2633
XM_017007781.1
XP_016863270.1
15, 16
2628
XM_017007782.1
XP_016863271.1
14, 15
2559
XM_017007783.1
XP_016863272.1
15, 16
2529
GenBank mRNA
AK092528.1
-
7, 8
889
AK128493.1
-
7, 8
1508
AK291025.1
-
7, 8
825
AK294476.1
-
14, 15
2577
AL133047.1
-
5, 6
501
BC010110.2
-
7, 8
930
BC085613.1
-
6, 7
716
BC108890.1
-
8, 9
1767
40001764
BC108891.1
-
8, 9
1767
40001765
BC108892.1
-
8, 9
1767
40001766
BC108893.1
-
7, 8
1659
40001767
BX647422.1
-
15, 16
2649
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
cytoskeleton organization
regulation of cell morphogenesis
positive regulation of membrane protein ectodomain proteolysis
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-wzvj9:80/10.239.161.253:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0